Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-DYKDDDDK Tag Mouse Monoclonal Antibody [clone: IE6]

Supplier: BioVision

3022-100
10003-228EA 252.35 USD InStock
10003-228
Anti-DYKDDDDK Tag Mouse Monoclonal Antibody [clone: IE6]
Antibodies
The DYKDDDDK Tag Antibody is a high-affinity mouse monoclonal antibody that can be used to detect DYKDDDDK-tagged proteins. It can detect N-terminal, C-terminal and internal tagged fusion proteins. This antibody recognizes the same epitope recognized by Sigma’s anti-FLAG® antibodies.

Recommended Dilutions: Western Blot: 0.5 µg/ml).

Type: Primary
Antigen: DYKDDDDK Tag
Clonality: Monoclonal
Clone: IE6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: All Species

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty