Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-AMY1A Rabbit Polyclonal Antibody

Supplier: BioVision

3014-100
10003-214EA 262.65 USD InStock
10003-214
Anti-AMY1A Rabbit Polyclonal Antibody
Antibodies
α-Amylase is an enzyme that is normally produced in the saliva glands and pancreas in humans and in other mammals. α-Amylase catalyzes the cleavage of 1, 4-alpha-D-glucosidic bonds of large sugar molecules into oligosaccharides and polysaccharides.

Recommended Dilutions: Western Blot: 1-4 µg/ml

Type: Primary
Antigen: AMY1A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty