Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-RELA Mouse Monoclonal Antibody [clone: 2A12A7]

Supplier: BioVision

3012-100
10003-212EA 350.2 USD InStock
10003-212
Anti-RELA Mouse Monoclonal Antibody [clone: 2A12A7]
Antibodies
NFkB was identified as a sequence specific transcriptional activator that binds to the intronic enhancer of kappa light chain gene in B lymphocytes. NFkB is a heterodimer that consists of a 50 kDa DNA binding subunit (p50) and a 65 kDa transactivation subunit (p65/RelA). Both of these subunits exhibit sequence homology to the protooncogene c-Rel. The p50/p65 heterodimer remains in the cytosol in an inactive form as a complex with its inhibitor, IkB. Upon stimulation of cells by a wide variety of stimuli such as lipopolysaccharide (LPS), pro-inflammatory cytokines, and viral infection. IkB is phosphorylated and degraded by proteosome. The active NFkB heterodimer is translocated into the nucleus and induces gene expression.

Recommended Dilutions: Western Blot: 1-4 µg/ml

Type: Primary
Antigen: RELA
Clonality: Monoclonal
Clone: 2A12A7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty