Anti-JUN Rabbit Polyclonal Antibody

Supplier: BioVision
3009-100
10003-204EA 286.04 USD
10003-204
Anti-JUN Rabbit Polyclonal Antibody
c-Jun is a component of the transcription factor AP-1 that binds and activates transcription at TRE/AP-1 elements. The transcriptional activity of c-Jun is regulated by phosphorylation at Ser63 and Ser73. Extracellular signals including growth factors, transforming oncoproteins, and UV irradiation stimulate phosphorylation of c-Jun at Ser63/73 and activate c-Jun dependent transcription. The MAP kinase homologue, JNK, binds to the N-terminal region of c-Jun and phosphorylates c-Jun at Ser63/73. The activity of JNK is stimulated by the same signals that activate c-Jun.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: JUN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR