Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-FADD Mouse Monoclonal Antibody [clone: T1B]

Supplier: BioVision

3008-100
10003-202EA 303.85 USD InStock
10003-202
Anti-FADD Mouse Monoclonal Antibody [clone: T1B]
Antibodies
Fas-associating death domain containing protein (FADD), also known as mediator of receptor induced toxicity (MORT1), is a 24 kDa cytosolic protein. The protein is involved in both Fas and TNF-R induced cell death pathway. FADD specifically binds to Fas, an association mediated by their homologous death domains. Overexpression of FADD induces apoptosis that can be inhibited by CrmA, a poxvirus protein that blocks both Fas- and TNF-induced cell death.

Recommended Dilutions: Western Blot: 1-4 µg/ml

Type: Primary
Antigen: FADD
Clonality: Monoclonal
Clone: T1B
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Dog

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty