Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-C-Peptide Mouse Monoclonal Antibody [clone: HCP-B2]

Supplier: BioVision

3103-100
10005-526EA 288.4 USD InStock
10005-526
Anti-C-Peptide Mouse Monoclonal Antibody [clone: HCP-B2]
Antibodies
C Peptide is part of the molecule of Proinsulin, the insulin precursor molecule, that consists of three parts: C Peptide and two long strands of amino acids (called the alpha and beta chains) that later become linked together to form the insulin molecule. From every molecule of proinsulin, one molecule of insulin plus one molecule of C Peptide are produced. C peptide is released into the blood stream in equal amounts to insulin. A test of C peptide levels will show how much insulin the body is making. The ratio of C-peptide and Proinsulin in human serum is also very important in diagnosis and prognosis of various diseases like Polycystic Ovary Syndrome, Ovarian carcinoma, etc.

Recommended Dilutions: indirect ELISA and IHC.

Type: Primary
Antigen: C-Peptide
Clonality: Monoclonal
Clone: HCP-B2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty