Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Amylase pancreatic Mouse Monoclonal Antibody [clone: RPA-B5]

Supplier: BioVision

3102-100
10005-524EA 272.95 USD InStock
10005-524
Anti-Amylase pancreatic Mouse Monoclonal Antibody [clone: RPA-B5]
Antibodies
Pancreatic amylase is a digestive enzyme secreted by pancreas into the intestine. It consists of a single polypeptide chain with a molecular weight of 54 kDa. Measurement of amylase concentrations in serum is a diagnostic test for various pancreatic disorders.

Recommended Dilutions: indirect ELISA and WB.

Type: Primary
Antigen: Amylase pancreatic
Clonality: Monoclonal
Clone: RPA-B5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty