Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-MAPKAPK2 Rabbit Polyclonal Antibody

Supplier: BioVision

3100-100
10005-516EA 272.95 USD InStock
10005-516
Anti-MAPKAPK2 Rabbit Polyclonal Antibody
Antibodies
MAP kinase-activated protein kinase-2 (MAPKAPK-2), also known as p45 hsp27 kinase, is a 45-54 kDa serine-threonine protein kinase that contains a proline rich sequence and two putative SH3-binding sites. MAPKAPK-2 is activated in response to stress, IL-1 and TNF, possibly catalyzed by p38/Hog-dependent phosphorylation. One of the major substrates of MAPKAPK-2 is hsp27, which stimulates actin polymerization in order to facilitate recovery from destruction of cytoskeleton during cellular stresses. Two isoforms are produced due to alternative splicing of the same gene which differs in their C-terminals. This antibody recognizes both isoforms.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml;

Type: Primary
Antigen: MAPKAPK2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Dog, Bovine

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty