Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-HSP70 Rabbit Polyclonal Antibody

Supplier: BioVision

3097-100
10003-368EA 252.35 USD InStock
10003-368
Anti-HSP70 Rabbit Polyclonal Antibody
Antibodies
Hsp70, one class of stress proteins, is comprised of multiple members, all of which bind ATP in vitro, but are localized within different intracellular compartments. These include: a) the constitutive Hsc70 (or cognate) present within the cytosol/nucleus; b) the highly stress-inducible Hsp70 present within the cytosol/ nucleus/nucleolus; c) the constitutive glucose regulated 78 kDa (or BiP) protein present within the lumen of the endoplasmic reticulum; and d) the glucose regulated 75 kDa protein present within the mitochondria. Hsp70 is typically not expressed in the cell under normal growth conditions, but is expressed at high levels in the cell experiencing stress. Consequently, detection of Hsp70 using an antibody specific for the inducible form is quite useful in ascertaining whether a stress response has occurred in the cell. It should be noted, however, that there is a constitutive level of Hsp70 expression in some cell types such as primates, e.g., human and monkey. Nevertheless increased expression of Hsp70 is observed in such cells following stress.

Recommended Dilutions: Western Blot: 1-2 µg/ml

Type: Primary
Antigen: HSP70
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Monkey, Hamster, Rabbit, Bovine

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty