Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-HSPA8 Rabbit Polyclonal Antibody

Supplier: BioVision

3095-100
10003-362EA 288.4 USD InStock
10003-362
Anti-HSPA8 Rabbit Polyclonal Antibody
Antibodies
The 70 kDa heat shock cognate protein, Hsc70, is part of the Hsp70 family. The family includes both cognate members and highly inducible isoforms. Members of the Hsp70 family are molecular chaperones which are involved in many cellular functions such as protein folding, transport, maturation and degradation.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml; in Immunohistochemistry

Type: Primary
Antigen: HSPA8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Monkey, Hamster, Rabbit, Bovine

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty