Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-HSPD1 Rabbit Polyclonal Antibody

Supplier: BioVision

3094R-100
10003-358EA 252.35 USD InStock
10003-358
Anti-HSPD1 Rabbit Polyclonal Antibody
Antibodies
Hsp60, also known as Cpn60, is an abundant protein synthesized constitutively in the cell that is induced to a higher concentration after brief cell stress or shock. It is present in all species analyzed so far and exhibits a remarkable sequence homology among various counterparts in bacteria, plants, and mammals with more than half of the residues identical between bacterial and mammalian Hsp60.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: HSPD1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty