Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-HSPB1 Rabbit Polyclonal Antibody

Supplier: BioVision

3093-100
10003-354EA 252.35 USD InStock
10003-354
Anti-HSPB1 Rabbit Polyclonal Antibody
Antibodies
Hsp27 exhibits a chaperone-like activity by serving as a site where unfolding proteins may bind until ATP and Hsp70-dependent refolding can occur. Hsp27 is believed to protect cells by enhancing cellular glutathione levels and elevated glutathione levels have been measured in cells overexpressing Hsp27. Data from studies using wild-type Hsp27 and mutant forms in which the serine phosphorylation sites were mutated to alanines, glycines or aspartates, have shown that cellular glutathione levels depend on the oligomerization of Hsp27. Recent findings indicate that Hsp27 is also a negative regulator of cytochrome c-dependent activation of procaspase-3.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: HSPB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty