Anti-LGALS1 Rabbit Polyclonal Antibody

Supplier: BioVision
3596-100
10004-340EA 286.04 USD
10004-340
Anti-LGALS1 Rabbit Polyclonal Antibody
Galectins are a family of soluble β-galactoside-binding animal lectins that modulate cell-to-cell and cell-to-ECM interactions, as well as apoptosis. Galectin-1 has the ability to induce apoptosis of activated T-cells and T-leukaemia cell lines. Other activities include cell differentiation and inhibition activity of CD45 protein phosphatase activity. Galectin also binds beta-galactoside as well as CD45, CD3 and CD4. Human Galectin is a 14.5 kDa protein consisting of 134 amino acid residues.

Recommended Dilutions: Western Blot: 0.5-2.0 µg/ml; Immunohistochemistry: 5 µg/ml

Type: Primary
Antigen: LGALS1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR