Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PLTP Rabbit Polyclonal Antibody

Supplier: BioVision

3595-100
10004-336EA 262.65 USD InStock
10004-336
Anti-PLTP Rabbit Polyclonal Antibody
Antibodies
Plasma phospholipid transfer protein (PLTP) plays a central role in regulating plasma HDL levels and distribution of HDL subpopulations. This regulation is evidenced in vivo by markedly reduced plasma HDL levels in PLTP gene knock-out mice. PLTP also facilitates free cholesterol influx into HDL.

Recommended Dilutions: Western Blot: 1:200-1000 dilutions; in Immunohistochemistry: 1:50 dilutions

Type: Primary
Antigen: PLTP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty