Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-VDAC Rabbit Polyclonal Antibody

Supplier: BioVision

3594-100
10004-332EA 288.4 USD InStock
10004-332
Anti-VDAC Rabbit Polyclonal Antibody
Antibodies
The Voltage-Dependent Anion Channel (VDAC or mitochondrial Porin)I is an outer membrane mitochondrial protein. The VDAC protein is thoμght to form the major pores throμgh which adenine nucleotides are transferred throμgh the outer mitochondrial membrane. VDAC has also been implicated in the formation of the mitochondrial permeability transition pore complex in apoptotic cells. This complex, formed by VDAC, ANT, and CypD is thoμght to allow the mitochondria to undergo metabolic uncoupling and irreversible morphologic changes that ultimately destroy the mitochondria during apoptosis.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml; Immunohistochemistry: 2.5 µg/ml

Type: Primary
Antigen: VDAC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Bovine, Rabbit

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty