Anti-DIO1 Rabbit Polyclonal Antibody

Supplier: BioVision
3592-100
10004-324EA 286.04 USD
10004-324
Anti-DIO1 Rabbit Polyclonal Antibody
Apoptosis plays an important role in many cellular processes. Dio-1 (Death Inducer-O bliterator-1), a putative transcription factor, has been indicated to function in the onset of apoptosis. Dio-1 expression is upregulated when an apoptosis is detected. It translocates to the nucleus, and activates apoptosis during limb development. Dio-1 expression can be suppressed by caspase inhibitors and Bcl-2 expression.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: DIO1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR