Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-LGALS3 Rabbit Polyclonal Antibody

Supplier: BioVision

3591-100
10004-320EA 252.35 USD InStock
10004-320
Anti-LGALS3 Rabbit Polyclonal Antibody
Antibodies
Galectins are a family of soluble β-galactoside-binding animal lectins that modulate cell-to-cell and cell-to-ECM interactions, as well as apoptosis. Galectin-3 encodes a 30-35 kDa protein containing a single carbohydrate binding domain, which binds galactose-containing glycoconjμgates. Upregulated expression of Galectin-3 is involved in cancer progression and metastasis.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: LGALS3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty