Anti-KISS1 Rabbit Polyclonal Antibody

Supplier: BioVision
AB286128-100UG
10004-316EA 1072.54 USD
10004-316
Anti-KISS1 Rabbit Polyclonal Antibody
Antibodies
KiSS-1 has been reported to suppress metastases of melanomas and breast carcinomas without affecting tumorigenicity. Human KiSS-1 transcripts are predominately expressed in the brain and placenta. Expression of KiSS-1 reduces the level of NFkB p50/p65 binding to the MMP9 promoter and correlates with diminished expression of the 92 kDa type IV collagenase (MMP-9). KiSS-1 may regulate events downstream of cell-matrix adhesion in mechanisms involving cytoskeletal reorganization.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: KISS1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR