Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-JNK1 Rabbit Polyclonal Antibody

Supplier: BioVision

3589-100
10004-314EA 334.75 USD InStock
10004-314
Anti-JNK1 Rabbit Polyclonal Antibody
Antibodies
The JNK pathway is activated by largely distinct stimuli including inflammatory cytokines (such as TNF-α and IL-1), UV light, inhibitors of protein synthesis and osmotic stress. Activated MEKK1 phosphorylates SEK1 (also known as MKK4), which in turn activates SAPK (also known as JNK). JNK binds tightly to the N-terminal region of c-Jun and ATF-2, and phosphorylates c-Jun at Ser63 and Ser73 and ATF-2 at Thr69 and Thr71.

Recommended Dilutions: Western Blot: 1-2 µg/ml; Immunoprecipitation: 10-20 µg/ml; Immunocytochemistry: 10-20 µg/ml; and Immunohistochemistry: 20-30 µg/ml

Type: Primary
Antigen: JNK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty