Anti-DMD Rabbit Polyclonal Antibody

Supplier: BioVision
3586R-100
10005-696EA 286.04 USD
10005-696
Anti-DMD Rabbit Polyclonal Antibody
Dystrophin is one of the actin-binding proteins that are involved in anchoring the cytoskeleton to the plasma membrane. Dystrophin expression is found in muscle brain tissues, where it is located to the inner surface of the plasma membrane. It is sμggested that alternative splicing of the caboxy terminus allows dystrophin to interact with a variety of proteins. Loss of dystrophin-associated proteins in Duchenne afflicted muscle is due to the absence of dystrophin rather than to muscle degradation and lack of dystrophin results in the loss of linkage between the cytoskeleton and extracellular matrix.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: DMD
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR