Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-LIMK2 Rabbit Polyclonal Antibody

Supplier: BioVision

3584-100
10005-688EA 272.95 USD InStock
10005-688
Anti-LIMK2 Rabbit Polyclonal Antibody
Antibodies
LIM kinases (LIMK1 and LIMK2) are serine/threnine kinases that have two zinc finger motifs, known as LIM motifs in their N-terminal regulatory domain. LIM kinases are involved in actin cytoskeleton regulation throμgh Rho-family GTPases and downstream kinases PAKs and ROCK. PAK1 and ROCK phosphorylate LIMK1 and LIMK2, which increases the activity of the kinases. Activated LIM kinases inhibit the actin depolymerization activity of cofilin by phosphorylation at the N-terminus of Cofilin.

Recommended Dilutions: Western Blot: 1-4 µg/ml

Type: Primary
Antigen: LIMK2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty