Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IRAK4 Rabbit Polyclonal Antibody

Supplier: BioVision

3580-100
10005-686EA 272.95 USD InStock
10005-686
Anti-IRAK4 Rabbit Polyclonal Antibody
Antibodies
The IRAK family members play an important role in IL-1R/TLR mediated inflammatory responses and in innate immunity. IRAK-4 is a novel member in the IRAK family. Over expression of IRAK-4 activates NFkB and MAPK pathways. Animas and human lacking IRAK-4 are impaired in their responses to viral and bacterial challenges.

Recommended Dilutions: Western Blot: 2-4 µg/ml

Type: Primary
Antigen: IRAK4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty