Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TNFRSF13C Rabbit Polyclonal Antibody

Supplier: BioVision

3578R-100
10005-682EA 252.35 USD InStock
10005-682
Anti-TNFRSF13C Rabbit Polyclonal Antibody
Antibodies
Members in the TNF superfamily regulate immune responses and induce apoptosis. A novel member in the TNF family was recently identified by several groups and designated BAFF, BLyS, TALL-1, THANK, and zTNF4. BAFF/BLyS was characterized as a B cell activator since it induced B cell proliferation and immunoglobulin secretion. Two receptors, TACI and BCMA, for BAFF were originally identified. A third receptor was identified recently and designated BAFF-R and BR3 for BLyS receptor 3. Unlike BCMA and TACI, which bind to BAFF and April, BAFF-R/BR3 is specific for BAFF and plays a predominant role in BAFF induced B cell development and survival. BAFF and its receptors are involved in B cell associated autoimmune diseases, and activate NF-kB and c-Jun N-terminal kinase.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: TNFRSF13C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty