Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TNFRSF17 Rabbit Polyclonal Antibody

Supplier: BioVision

3577-100
10005-680EA 272.95 USD InStock
10005-680
Anti-TNFRSF17 Rabbit Polyclonal Antibody
Antibodies
BCMA is a recently identified receptor for BAFF. It binds BAFF and April. BAFF and its receptors induce B cell development and survival, activate NF-kB and c-Jun N-terminal kinase, and are involved in B cell associated autoimmune diseases.

Recommended Dilutions: Western Blot: 2-6 µg/ml; Immunohistochemistry: 5-20 µg/ml

Type: Primary
Antigen: TNFRSF17
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty