Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-WNT2 Rabbit Polyclonal Antibody

Supplier: BioVision

3572R-100
10005-666EA 288.4 USD InStock
10005-666
Anti-WNT2 Rabbit Polyclonal Antibody
Antibodies
Wnt gene family members, including Wnt-1 throμgh Wnt-10, play a key role in regulating cellular growth and differentiation. Wnt-1 functions as a key regulator of cellular adhesion. Wnt-3 is also involved in tumorigenesis and Wnt-2 and Wnt-4 may be associated with abnormal proliferation in human breast tissue. Wnt-1, Wnt-3 and Wnt-10b have been implicated along with FGF-3 in the development of mouse mammary tumor virus induced mouse mammary carcinomas.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: WNT2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty