Would you like to log in again?
m-TGE Media with Indicator, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS
TCI200 TCI200
10803-106PK 129.77 USD InStock
CA10803-106 10803-106
m-TGE Media with Indicator, Hardy Diagnostics
Media for Microbiology Additives and Supplements for Microbiology
Used with membrane filters to culture bacteria, yeasts, and molds
Quality certificates are included with each shipment, and packages are clearly marked with expiration date.
Available ready-to-use 2ml ampoules contain enough medium to saturate one standard absorbent pad in a petri dish; 50 ampoules per box. Single use vials assure sterility. Each product is lot tested and certified to comply with established test criteria.
Ordering information: Media is packaged 50 ampoules per pack.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty