Would you like to log in again?
Thumbs-Up Microtubes, Diversified Biotech
Supplier: DIVERSIFIED BIOTECH
THUM-2100 THUM-2500 THUM-2000 THUM-2400 THUM-2300 THUM-2200
10141-840PK 35.26 USD InStock
10141-840 490007-004 490006-998 490007-002 490007-000 10141-842
Thumbs-Up Microtubes, Diversified Biotech
Tubes Microtubes
The tube with the thumb tab
  • Using thumb tab for tube closure prevents closing gloves under lid
  • Thumb tab allows for quick removal of your tube from crowded racks
  • Upward angle of thumb tab creates a smaller circumference which allows Thumbs-Up tubes to fit more easily next to one another in tube racks
  • RNase, DNase and Pyrogen free
  • -196°C (vapor phase only) to 130°C
  • RCF rating of 22,000 Xg
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty