Would you like to log in again?
Kimberly-Clark® Purple Nitrile® Sterile Exam Gloves, Singles, Kimberly-Clark Professional®
Supplier: KIMBERLY CLARK CORP
KIMBERLY-CLARK®
52103 52101 52102
10141-356CS 511.81 USD InStock
10141-356 10141-352 10141-354
Kimberly-Clark® Purple Nitrile® Sterile Exam Gloves, Singles, Kimberly-Clark Professional®
Gloves
Kimberly-Clark Purple Nitrile Exam Gloves are made of a highly durable nitrile polymer that eliminates the risks and costs associated with type 1 natural rubber latex allergies
Comfortable to wear and provide excellent dexterity and tactile sensitivity, plus textured fingertips enhance wet and dry grip.
Certifications: Cleared for use in chemotherapy. Manufactured in accordance with Quality System ISO 9002. Exceeds current ASTM 6319 standard for critical defects (AQL 2.5). AQL for critical defects is 1.5.
Ordering information: Gloves are eligible for recycling with Kimberly-Clark Professional RightCycle Program. Gloves are packaged 100 gloves per box with 4 boxes per case.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty