DuraSeal™ Cling, Diversified Biotech
DSC1-150
490014-304EA 42.07 USD
490014-304
DuraSeal™ Cling, Diversified Biotech
  • Film can cling to itself and glass vessels
  • Softens at 100°C
  • Film can stretch 300-400% before breaking or tearing
  • Forms an air tight barrier when stretched across openings