Would you like to log in again?
Electropolished Stainless Steel Freezer Racks, Diversified Biotech
Supplier: DIVERSIFIED BIOTECH
SSFR-6000 SSFR-5000 SSFR-2000 SSFR-4000 SSFR-7000 SSFR-8000 SSFR-1000 SSFR-3000
10141-938EA 216.94 USD InStock
10141-938 10141-936 10141-832 10141-934 10141-834 10141-836 10141-830 10141-932
Electropolished Stainless Steel Freezer Racks, Diversified Biotech
Racks Test Tube Racks
High-quality, ultra-durable freezer racks for long term sample storage
  • All edges have been finished to remove the sharp surfaces that plague standard freezer racks
  • Electropolishing cleans up weld points that may be the starting points for corrosion
  • Superior construction and design creates a more sturdy rack
  • Designed and finished to last longer in harsh freezer environment
  • Racks are ultra-shiny and free from any surface contaminants
  • Vertical and horizontal options are available to fit a variety of freezer configurations
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty