Silver BioGuard Anti-Microbial Mats
SLVR-3200 SLVR-3000 SLVR-3100
10141-802EA 39.3 USD
10141-802 10141-800 10141-798
Silver BioGuard Anti-Microbial Mats
Silver BioGuard anti-microbial mats minimize workspace contamination
Mats are less than 1mm thick and are anti-skid making them perfect for the benchtop or under the hood.