PetriDish Support-N-Seal, Diversified Biotech
Supplier: Diversified Biotech
PDHL-1000
10141-852EA 61.02 USD
10141-852
PetriDish Support-N-Seal, Diversified Biotech
Racks Petri Dish Racks
Support-N-Seal provides a stabilizing vertical slot in which the Petri dish and lid are held closely together while allowing rotation as a sealing tape is applied around the dish.
Compatible with most sealing tapes. A wide base and non-slip feet ensure that the Support-N-Seal remains in place while in use. Fits Petri dishes up to 100mm in diameter and 15mm high (including lid).