Would you like to log in again?
Gel Handler, Diversified Biotech
Supplier: DIVERSIFIED BIOTECH
GH-100
490006-750EA 44.19 USD InStock
490006-750
Gel Handler, Diversified Biotech
Electrophoresis Plates and Sheets
Flexible UV transparent sheets for handling gels, visualizing and photo-documentation.
  • Thin sheet that easily slides beneath a gel
  • Flexibility of the sheet eliminates air bubbles
  • Protects the transilluminator from cuts and scratches
  • Polyolefin composition allows acid washing and autoclaving without a chemical reaction
Optical densities for the 5 mil thick sheets at 254 and 300 nm emission wave lengths of the transilluminator are 0.14 and 0.04 respectively.
Packaging: Sheets are packaged 10 per pack.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty