ChillDown Agarose and Chemical Cooler, Diversified Biotech
CHDN-1000
10141-848EA 91.69 USD
10141-848
ChillDown Agarose and Chemical Cooler, Diversified Biotech
  • Chill molten agarose down to 60°C in minutes
  • Rapid cooling of agarose prevents an early-polymerized layer on top of the solution, which interferes with results
  • Use ChillDown whenever cooling of solutions is desirable
  • Fits any 250ml beaker
  • Leave ChillDown in the freezer overnight and it will be ready for you when needed
  • Polyethylene beaker cooler filled with propylene glycol