Would you like to log in again?
Staticmaster® Auto-Balanced Ionizing Blowers, NRD
Supplier: NRD Staticmaster
6212 6211
75794-888EA 523.66 USD InStock
75794-888 75794-886
Staticmaster® Auto-Balanced Ionizing Blowers, NRD
Ionizers
The Staticmaster® 6211 and 6212 Auto-Balanced Ionizing Blowers utilize NRD's auto­balancing technology, making them suitable for semiconductor back-end, printed circuit board assembly, and general electronics applications
  • Auto-balance technology
  • Facility Monitoring System (FMS) interface
  • LED alarm indiators
  • No calibration required
Neither model requires periodic calbration to maintain its low offset voltage balance. The Facility Monitoring System (FMS) interface wlll allow real time Ionizer status monitoring and provides visual and audio alarms for ionizer malfunctions or power down. The Model 6212 is slightly larger than Model 6211 with greater airflow.
Order Now
ORDER
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty