Hafnium ≥99.8% (trace metals basis excluding Zr), powder -100 mesh (Zr ∼ 4.5%)

Danger

77749-22
AA77749-22EA 209.5 USD
AA77749-22
Hafnium ≥99.8% (trace metals basis excluding Zr), powder -100 mesh (Zr ∼ 4.5%)
Hafnium
Formula: Hf
MW: 178.49 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011032
CAS Number: 7440-58-6
EINECS: 231-166-4
UN: 2545
ADR: 4.2,II

Order Now

Specification Test Results

Assay Minimum 99.8

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR