Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Dibismuth triselenide ≥99.995% (metals basis)

Supplier: Alfa Aesar
Danger
Synonyms: Bismuth (III) selenide

47198-06 47198-03
AA47198-06EA 119.64 USD InStock
AA47198-06 AA47198-03
Dibismuth triselenide ≥99.995% (metals basis)
Dibismuth triselenide
  • -325 Mesh Powder


Formula: Bi₂Se₃
MW: 654.84 g/mol
Melting Pt: 710 °C
Density: 6,82
Storage Temperature: Ambient
MDL Number: MFCD00014200
CAS Number: 12068-69-8
EINECS: 235-104-7
UN: 3283
ADR: 6.1,III
Merck Index: 12,01319
Order Now
Specification Test Results
Assay Minimum 99.999

 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty