Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

n-Heptane, anhydrous, over molecular sieves

Supplier: Alfa Aesar
Danger
Synonyms: DipropylmethaneEptaniHeptanHeptaneHeptanenHeptyl hydrideSkellysolve c

Packaged under Argon in resealable ChemSeal™ bottles.

ChemSeal
47191-K2 47191-AK
AA47191-K2EA 73.15 USD InStock
AA47191-K2 AA47191-AK
n-Heptane, anhydrous, over molecular sieves
n-Heptane
  • Liquid


Formula: H₃C(CH₂)₅CH₃
MW: 100.2 g/mol
Boiling Pt: 97-99 °C
Melting Pt: -91 °C
Density: 0,684
Flash Pt: -4 °C (24 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009544
CAS Number: 142-82-5
EINECS: 205-563-8
UN: 1206
ADR: 3,II
Merck Index: 13,04679
https://us.vwr.com/stibo/web/std.lang.all/64/15/736415.jpg
Order Now

 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty