Tantalum, gauze woven from wire 100 mesh woven from 0.08mm (0.003in)

Supplier: Thermo Scientific Chemicals

46695-FL 046695-FL 46695-GT 46695-GT
AA46695-FLEA 140.99 USD
CAAA46695-FL AA46695-FL CAAA46695-GT AA46695-GT
Tantalum, gauze woven from wire 100 mesh woven from 0.08mm (0.003in)
Tantalum

  • Wire Cloth

Formula: Ta
Storage Temperature: Ambient
MDL Number: MFCD00011252
CAS Number: 7440-25-7
EINECS: 231-13505

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR