Imide-extended bismaleimide, gel BMI-3000

Warning

46532-14 46532-14 46532-22 46532-22
AA46532-22EA 660.91 USD
AA46532-22 CAAA46532-22 AA46532-14 CAAA46532-14
Imide-extended bismaleimide, gel BMI-3000
Imide-extended bismaleimide

Gel

Storage Temperature: Refrigerator
CAS Number: 1290041-56-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR