Would you like to log in again?
Duplex 2209 alloy, wire 60 mesh, Diameter 0.15 mm (0.006 mm)
Supplier: Alfa Aesar
Danger
46525-GT 46525-FL 46525-GT 46525-FL
AA46525-GTEA 65.71 USD InStock
AA46525-GT AA46525-FL CAAA46525-GT CAAA46525-FL
Duplex 2209 alloy, wire 60 mesh, Diameter 0.15 mm (0.006 mm)
Duplex 2209 alloy
Duplex 2209, 60 mesh woven from 0.15mm (0.006mm) dia wire
Wire Cloth
Typical composition of alloys are Cr 21.5 to 23.5%, Ni 7 to 9%, Mo 2.5 to 3.5%, Mn 0.5 to 2%, Cu 0.75% max, balance Fe.
Order Now
ORDER
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty