Would you like to log in again?
Platinum ruthenium alloy (95:5 wt%), wire Ø 1.5 mm (0.059 in) x 150 mm (5.9 in) length, hard straight lengths
Supplier: Alfa Aesar
46428-KT 46428-KT
AA46428-KTEA 978.5 USD InStock
AA46428-KT CAAA46428-KT
Platinum ruthenium alloy (95:5 wt%), wire Ø 1.5 mm (0.059 in) x 150 mm (5.9 in) length, hard straight lengths
Platinum ruthenium alloy (95:5 wt%)
Pt:Ru; 95:5 wt%, ~5.48g/ea
Order Now
ORDER
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty