Ruthenium 5% on activated carbon powder wetted with ca. 50% water, reduced by hydrogen Type D101023-5

45439-06 45439-14
AA45439-14EA 158.73 USD
AA45439-14 AA45439-06
Ruthenium 5% on activated carbon powder wetted with ca. 50% water, reduced by hydrogen Type D101023-5
Ruthenium
Formula: Ru
MW: 101.07 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03458417
CAS Number: 7440-18-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR