Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

S5-Alcohol Dehydrogenase

Supplier: Alfa Aesar
45399-4I 45399-4I
AA45399-4IEA 114.59 USD InStock
CAAA45399-4I AA45399-4I
S5-Alcohol Dehydrogenase
Enzymes
Enzymic catalysis in organic solvents

Pale yellow lyophilized powder

CAS No.: 9031-72-5
Physical Form: Powder
Appearance: Pale yellow
MDL No.: MFCD00081305

Applications: Enzymic catalysis in organic solvents

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty