Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Lead in Isooctane standard solution, Specpure®, 3.70µg/g(0.010g/gal)

Supplier: Alfa Aesar
Specpure
45333-22 45333-22
AA45333-22EA 123.6 USD InStock
AA45333-22 CAAA45333-22
Lead in Isooctane standard solution, Specpure®, 3.70µg/g(0.010g/gal)
Standards Petroleum Industry Standards
Liquid

Molecular Formula: Matrix: Isooctane
Physical Form: Liquid
Appearance: Colorless
Odor: Nearly odorless
UN No.: 1262
MDL No.: MFCD01094200

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty