Polypropylene sheet, Thickness 6.36mm (0.25in)

Supplier: Thermo Scientific Chemicals

45174-HB 045174-HB
AA45174-HBEA 40.79 USD
AA45174-HB CAAA45174-HB
Polypropylene sheet, Thickness 6.36mm (0.25in)
Polypropylene sheet
Formula: C₂₂H₄₂O₃
MW: 354.575 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00084447
CAS Number: 9003-07-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR