Would you like to log in again?
Monel® 400 alloy, gauze woven from wire 40 mesh woven from 0.25mm (0.01in)
Supplier: Alfa Aesar
45163-GT 45163-HB 45163-GT 45163-HB
AA45163-GTEA 21.63 USD InStock
AA45163-GT AA45163-HB CAAA45163-GT CAAA45163-HB
Monel® 400 alloy, gauze woven from wire 40 mesh woven from 0.25mm (0.01in)
Monel® 400 alloy
Typical composition of alloy: Ni 63.0% min, Cu 28.0-34.0%, C 0.3% max, Mn 2.0% max, Fe 2.5% max, S 0.024% max, Si 0.5% max
Order Now
ORDER
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty