PTFE, rods Ø 3.18 mm (0.125 in)
Supplier: Thermo Scientific Chemicals
Synonyms:
Teflon
045152-BV
45152-BV
AA45152-BVEA
30.63
USD
AA45152-BV
CAAA45152-BV
PTFE, rods Ø 3.18 mm (0.125 in)
PTFE
Formula:
(CF₂CF₂)n Storage Temperature: Ambient |
MDL Number:
MFCD00084455 CAS Number: 9002-84-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...