Nichrome etchant
Supplier: Thermo Scientific Chemicals
|
Danger
|
44585-R0
44585-R0
044585-LV
44585-LV
AA44585-LVEA
452.99
USD
AA44585-LV
CAAA44585-R0
CAAA44585-LV
AA44585-R0
Nichrome etchant
Nichrome
Nichrome Etchant is a high purity system based on ceric ammonium nitrate. It provides clean, precise etching of evaporated nichrome layers for the preparation of thin film circuits. This product can be operated at room temperature or elevated temperature to increase etch rate.
Semiconductor-grade ingredients for ultra-high purity
Etchant filter to 0,2 microns to remove particles
Density:
1.25 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD07370976 UN: 3264 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...