Would you like to log in again?
POCO Electron Beam Crucibles, Alfa Aesar
Supplier: Alfa Aesar
38677-KT 38683-KT 38678-KT 38677-KT 41866-KT 38681-KT 38675-KT 38684-KT 38676-KT 38683-KT 40819-KT 41866-KT 38680-KT 38676-KT 38681-KT 40819-KT 38682-KT 43950-KT 38680-KT 38684-KT 38678-KT 38675-KT 43198-KT 38682-KT
AA38677-KTEA 122.15 USD InStock
CAAA38677-KT CAAA38683-KT CAAA38678-KT AA38677-KT AA41866-KT CAAA38681-KT AA38675-KT AA38684-KT AA38676-KT AA38683-KT CAAA40819-KT CAAA41866-KT CAAA38680-KT CAAA38676-KT AA38681-KT AA40819-KT AA38682-KT AA43950-KT AA38680-KT CAAA38684-KT AA38678-KT CAAA38675-KT AA43198-KT CAAA38682-KT
POCO Electron Beam Crucibles, Alfa Aesar
Crucibles
The POCO graphite electron beam evaporation crucibles are designed to offer E-Beam users improved evaporation performance over that obtainable in bare hearth mode
  • Ultrafine-grain
  • Isotropic microstructure
  • High density
  • Strong polycrystalline material
  • Inter-connecting porosity
  • Purified to below 5 ppm
These crucibles allow an increased deposition rate of up to 400% with the same power, or the same deposition using only 25% of the power, when compared to evaporation from a bare hearth. POCO crucibles can allow a much lower power setting while maintaining an acceptable deposition rate.
These graphite crucibles act as an “energy efficient” thermal barrier between the molten evaporant and the water cooled copper hearth.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty